Gene Transcripts

Gene Symbol
Zfp382
Entrez Gene ID
246264
Gene Name
zinc finger protein 382
Gene Aliases
Ks1, Znf382
Chromosome Location
Chr. 1: 88582641 - 88603219 on Build Rnor_6.0
UniGene ID
Rn.11374
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144749.1 NP_653350.1 1063
GenBank mRNA BC127483.1 - 476
U56732.1 - 1063

View Full Product Details