Gene Transcripts

Gene Symbol
Fxyd3
Entrez Gene ID
17178
Gene Name
FXYD domain-containing ion transport regulator 3
Gene Aliases
Mat8
Chromosome Location
Chr. 7: 31070535 - 31076697 on Build GRCm38
UniGene ID
Mm.263847
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008557.2 NP_032583.1 8 490
GenBank mRNA AK133713.1 - 1 1483
AK145206.1 - 461
AK166426.1 - 8 463
BC002039.1 - 8 454 3488180
BC051033.1 - 6 851 4460667
BC056223.1 - 8 458 6485767
EH101686.1 - 2 346
X93038.1 - 8 486

View Full Product Details