Gene Transcripts

Gene Symbol
Coil
Entrez Gene ID
12812
Gene Name
coilin
Gene Aliases
C79982, Cln80, p80, p80-coilin
Chromosome Location
Chr. 11: 88973935 - 88991613 on Build GRCm38
UniGene ID
Mm.268004
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF208663.1 - 1, 2 261

View Full Product Details