Gene Transcripts

Gene Symbol
Ppp6r2
Entrez Gene ID
71474
Gene Name
protein phosphatase 6, regulatory subunit 2
Gene Aliases
1110033O10Rik, 8430411H09Rik, B230107H12Rik, Pp6r2, Saps2
Chromosome Location
Chr. 15: 89211521 - 89287015 on Build GRCm38
UniGene ID
Mm.184055
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027805.2 NP_082081.1 8 1221
GenBank mRNA AK018403.1 - 8 1140
AK084684.1 - 8 1222

View Full Product Details