Gene Transcripts

Gene Symbol
Dab1
Entrez Gene ID
13131
Gene Name
disabled 1
Gene Aliases
AI956902, C630028C02Rik, scm, scr, scrambler, yot
Chromosome Location
Chr. 4: 103619546 - 104744844 on Build GRCm38
UniGene ID
Mm.289682
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177259.4 NP_796233.2 15 3858
GenBank mRNA AK045616.1 - 1 1165
AK083206.1 - 2 1922

View Full Product Details