gRNA Design Details
- Target DNA Sequence
- GGAAAAGATCGCAACGCCAA
- PAM Sequence
- AGG
- Target locus
-
Chr.7: 143267664 - 143267686 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
91.72/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GSTK1
- Entrez Gene ID
-
373156
- Gene Name
- glutathione S-transferase kappa 1
- Gene Aliases
-
GST, GST 13-13, GST13, GST13-13, GSTK1-1, hGSTK1
- Chromosome Location
-
Chr.7: 143263429 - 143269129 on GRCh38
- UniGene ID
- Hs.390667
- Species
-
Human