gRNA Design Details
- Target DNA Sequence
- GACACAGAAGTACAAGCGCC
- PAM Sequence
- AGG
- Target locus
-
Chr.6: 31271659 - 31271681 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
89.58/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- HLA-C
- Entrez Gene ID
-
3107
- Gene Name
- major histocompatibility complex, class I, C
- Gene Aliases
-
D6S204, HLA-JY3, HLAC, HLC-C, MHC, PSORS1
- Chromosome Location
-
Chr.6: 31268749 - 31272136 on GRCh38
- UniGene ID
- Hs.656020
- Species
-
Human