gRNA Design Details
- Target DNA Sequence
- TGGATGATTGGCTGCGACCT
- PAM Sequence
- GGG
- Target locus
-
Chr.6: 29828560 - 29828582 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
90.79/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- HLA-G
- Entrez Gene ID
-
3135
- Gene Name
- major histocompatibility complex, class I, G
- Gene Aliases
-
MHC-G
- Chromosome Location
-
Chr.6: 29826967 - 29831130 on GRCh38
- UniGene ID
- Hs.512152
- Species
-
Human