gRNA Design Details

Target DNA Sequence
TGGATGATTGGCTGCGACCT
PAM Sequence
GGG
Target locus
Chr.6: 29828560 - 29828582 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 90.79/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
HLA-G
Entrez Gene ID
3135
Gene Name
major histocompatibility complex, class I, G
Gene Aliases
MHC-G
Chromosome Location
Chr.6: 29826967 - 29831130 on GRCh38
UniGene ID
Hs.512152
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_002127.5 NP_002118.1 4
XM_017010816.1 XP_016866305.1 4