gRNA Design Details
- Target DNA Sequence
- CATCGAAGTATCTCAGGGTC
- PAM Sequence
- AGG
- Target locus
-
Chr.7: 18262503 - 18262525 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
88.31/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Mill1
- Entrez Gene ID
-
266815
- Gene Name
- MHC I like leukocyte 1
- Gene Aliases
-
5530400I18Rik
- Chromosome Location
-
Chr.7: 18245232 - 18266092 on GRCm38
- UniGene ID
- Mm.106769
- Species
-
Mouse