gRNA Design Details
- Target DNA Sequence
- GAAAATCTGTGCGGTGTCCC
- PAM Sequence
- TGG
- Target locus
-
Chr.6: 30489924 - 30489946 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
88.65/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- HLA-E
- Entrez Gene ID
-
3133
- Gene Name
- major histocompatibility complex, class I, E
- Gene Aliases
-
EA1.2, EA2.1, HLA-6.2, MHC, QA1
- Chromosome Location
-
Chr.6: 30489406 - 30494205 on GRCh38
- UniGene ID
- Hs.650174
- Species
-
Human