gRNA Design Details

Target DNA Sequence
TGCTCGTAAATTTGCCAGAG
PAM Sequence
AGG
Target locus
Chr.1: 75732678 - 75732700 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 90.37/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
ACADM
Entrez Gene ID
34
Gene Name
acyl-CoA dehydrogenase, C-4 to C-12 straight chain
Gene Aliases
ACAD1, MCAD, MCADH
Chromosome Location
Chr.1: 75724347 - 75763679 on GRCh38
UniGene ID
Hs.445040
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_000016.5 NP_000007.1 3
NM_001127328.2 NP_001120800.1 3
NM_001286042.1 NP_001272971.1 2
NM_001286043.1 NP_001272972.1 3
NM_001286044.1 NP_001272973.1 2