Gene Transcripts
- Gene Symbol
- STRADB
- Entrez Gene ID
- 55437
- Gene Name
- STE20-related kinase adaptor beta
- Gene Aliases
- ALS2CR2, CALS-21, ILPIP, ILPIPA, PAPK, PRO1038
- Chromosome Location
- Chr. 2: 201451669 - 201480851 on Build GRCh38
- UniGene ID
- Hs.652338
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001206864.1 |
NP_001193793.1
|
8, 9 |
1089 |
|
NM_018571.5 |
NP_061041.2
|
8, 9 |
1089 |
|
XM_005246669.1 |
XP_005246726.1
|
8, 9 |
1043 |
|
XM_017004438.1 |
XP_016859927.1
|
7, 8 |
924 |
|
GenBank mRNA |
AB038950.1 |
- |
8, 9 |
965 |
|
AF116618.2 |
- |
4, 5 |
526 |
|
AK024720.1 |
- |
4, 5 |
443 |
|
AK027637.1 |
- |
8, 9 |
1089 |
|
AK290457.1 |
- |
9, 10 |
953 |
|
AY093697.1 |
- |
8, 9 |
977 |
|
BC008302.1 |
- |
8, 9 |
991 |
3546243 |
BC090871.1 |
- |
8, 9 |
962 |
30530122 |
View Full Product Details