Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020309.3 | NP_064705.1 | 4 | 630 | |
GenBank mRNA | AB032436.1 | - | 4 | 479 | |
AK057879.1 | - | 576 | |||
AK095659.2 | - | 3 | 1909 | ||
AK289622.1 | - | 1 | 38 | ||
AK294226.1 | - | 4 | 436 | ||
AK294405.1 | - | 4 | 479 | ||
BC059379.1 | - | 4 | 605 | 5262415 | |
BG701511.1 | - | 4 | 607 |