Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 251327-251600 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSMD13
Entrez Gene ID
5719
Gene Name
proteasome 26S subunit, non-ATPase 13
Gene Aliases
HSPC027, Rpn9, S11, p40.5
UniGene
Hs.134688
Gene Chromosome Location
Chr.11: 236675-252984 on Build GRCh38
Transcripts
NM_002817.3 NM_175932.2 XM_011520235.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6445643 c.846G>A p.M282I Chr.11: 251548-251548 on Build GRCh38
314533 c.849A>T p.T283T Chr.11: 251557-251557 on Build GRCh38
314534 c.855A>T p.T285T Chr.11: 251557-251557 on Build GRCh38
4032151 c.876A>G p.Q292Q Chr.11: 251578-251578 on Build GRCh38
6445644 c.840G>A p.M280I Chr.11: 251548-251548 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs976683817 Chr.11: 251474-251474 on Build GRCh38
rs772100804 Chr.11: 251519-251519 on Build GRCh38
rs139250939 Chr.11: 251481-251481 on Build GRCh38
rs776493718 Chr.11: 251557-251557 on Build GRCh38
rs1027041083 Chr.11: 251381-251381 on Build GRCh38
rs551073302 Chr.11: 251421-251421 on Build GRCh38
rs925667918 Chr.11: 251568-251568 on Build GRCh38
rs1018115199 Chr.11: 251409-251409 on Build GRCh38
rs35322057 Chr.11: 251426-251426 on Build GRCh38
rs374743572 Chr.11: 251533-251533 on Build GRCh38
rs866942707 Chr.11: 251433-251433 on Build GRCh38
rs1044561055 Chr.11: 251390-251390 on Build GRCh38
rs1021673603 Chr.11: 251528-251528 on Build GRCh38
rs757341450 Chr.11: 251497-251497 on Build GRCh38
rs760680640 Chr.11: 251547-251547 on Build GRCh38
rs200386830 Chr.11: 251535-251535 on Build GRCh38
rs1006687597 Chr.11: 251402-251402 on Build GRCh38
rs750396962 Chr.11: 251466-251466 on Build GRCh38
rs747723527 Chr.11: 251502-251502 on Build GRCh38
rs780135441 Chr.11: 251476-251476 on Build GRCh38
rs952529991 Chr.11: 251388-251388 on Build GRCh38
rs994679914 Chr.11: 251354-251354 on Build GRCh38
rs907433773 Chr.11: 251392-251392 on Build GRCh38
rs766051485 Chr.11: 251550-251550 on Build GRCh38
rs1031613106 Chr.11: 251473-251473 on Build GRCh38

View Full Product Details