Gene Transcripts
- Gene Symbol
- Slc13a3
- Entrez Gene ID
- 114644
- Gene Name
- solute carrier family 13 (sodium-dependent dicarboxylate transporter), member 3
- Gene Aliases
- NaDC-3, NaDC3, SDCT2
- Chromosome Location
- Chr. 2: 165405295 - 165473225 on Build GRCm38
- UniGene ID
- Mm.250738
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_054055.2 |
NP_473396.1
|
1, 2 |
156 |
|
GenBank mRNA |
AF306491.1 |
- |
1, 2 |
155 |
|
AK085326.1 |
- |
1, 2 |
154 |
|
AK086775.1 |
- |
1, 2 |
158 |
|
AK087518.1 |
- |
1, 2 |
154 |
|
AK128982.1 |
- |
1, 2 |
153 |
|
AK138344.1 |
- |
1, 2 |
154 |
|
AK157881.1 |
- |
1, 2 |
125 |
|
AK165355.1 |
- |
1, 2 |
154 |
|
BC026803.1 |
- |
1, 2 |
142 |
4238769 |
BC028882.1 |
- |
1, 2 |
140 |
|
View Full Product Details