Gene Transcripts

Gene Symbol
Clic5
Entrez Gene ID
224796
Gene Name
chloride intracellular channel 5
Gene Aliases
5730531E12Rik, B330005L24, Gm322, jbg
Chromosome Location
Chr. 17: 44134678 - 44280168 on Build GRCm38
UniGene ID
Mm.37666
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172621.2 NP_766209.1 6 982
GenBank mRNA AK017800.1 - 6 943
AK046522.1 - 6 971
AK156849.1 - 6 823
BC064037.1 - 6 967 1227040

View Full Product Details