Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025968.3 | NP_080244.1 | 8 | 770 | |
GenBank mRNA | AK010888.1 | - | 8 | 805 | |
AK011962.1 | - | 8 | 792 | ||
AK035425.1 | - | 8 | 771 | ||
AK134440.1 | - | 8 | 803 | ||
AK166835.1 | - | 8 | 770 | ||
BC014865.1 | - | 8 | 752 | 2648051 | |
CT010267.1 | - | 7 | 719 |