Gene Transcripts

Gene Symbol
Vav2
Entrez Gene ID
22325
Gene Name
vav 2 oncogene
Gene Aliases
2810040F13Rik, AI847175
Chromosome Location
Chr. 2: 27262104 - 27427608 on Build GRCm38
UniGene ID
Mm.179011
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009500.1 NP_033526.1 27, 28 2911
GenBank mRNA BC053060.1 - 29, 30 2988
U37017.1 - 27, 28 2911

View Full Product Details