Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001080979.1 | NP_001074448.2 | 9 | 1399 | |
NM_011567.2 | NP_035697.3 | 10 | 1528 | ||
GenBank mRNA | AB221623.1 | - | 8 | 829 | |
AK133915.1 | - | 10 | 1190 | ||
BC130257.1 | - | 8 | 834 | 40142595 | |
D87965.1 | - | 9 | 1399 | ||
D87966.1 | - | 10 | 1523 | ||
L26343.1 | - | 10 | 1068 | ||
L26344.1 | - | 1013 | |||
U51743.1 | - | 8 | 814 | ||
X94441.1 | - | 9 | 959 |