Gene Transcripts

Gene Symbol
Tmed3
Entrez Gene ID
66111
Gene Name
transmembrane emp24 domain containing 3
Gene Aliases
1200002G13Rik, AW546672, P24b
Chromosome Location
Chr. 9: 89699203 - 89705043 on Build GRCm38
UniGene ID
Mm.27606
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025360.2 NP_079636.2 1 154
GenBank mRNA AK004545.1 - 1 102
AK007544.1 - 1 96
BC023338.1 - 1 48 5253777
BY159164.1 - 1 154

View Full Product Details