Gene Transcripts
- Gene Symbol
- Uba7
- Entrez Gene ID
- 74153
- Gene Name
- ubiquitin-like modifier activating enzyme 7
- Gene Aliases
- 1300004C08Rik, Ube1l
- Chromosome Location
- Chr. 9: 107975567 - 107984056 on Build GRCm38
- UniGene ID
- Mm.277125
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_023738.4 |
NP_076227.1
|
10, 11 |
1300 |
|
GenBank mRNA |
AF288694.1 |
- |
5, 6 |
562 |
|
AK004894.2 |
- |
10, 11 |
1301 |
|
AK054410.1 |
- |
10, 11 |
1307 |
|
AK089235.1 |
- |
10, 11 |
1304 |
|
AK089256.1 |
- |
4, 5 |
732 |
|
AK170421.1 |
- |
10, 11 |
1293 |
|
AK170644.1 |
- |
9, 10 |
1675 |
|
AK172467.1 |
- |
9, 10 |
1638 |
|
BC037693.1 |
- |
8, 9 |
1074 |
|
BC138843.1 |
- |
10, 11 |
1217 |
8861865 |
View Full Product Details