Gene Transcripts

Gene Symbol
Mamdc4
Entrez Gene ID
252882
Gene Name
MAM domain containing 4
Gene Aliases
Aegp
Chromosome Location
Chr. 3: 2786523 - 2795109 on Build Rnor_6.0
UniGene ID
Rn.10059
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145768.1 NP_665711.1 16 1923
GenBank mRNA L37380.1 - 16 1923

View Full Product Details