Gene Transcripts

Gene Symbol
Crmp1
Entrez Gene ID
25415
Gene Name
collapsin response mediator protein 1
Gene Aliases
Chromosome Location
Chr. 14: 78266931 - 78312778 on Build Rnor_6.0
UniGene ID
Rn.11379
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012932.2 NP_037064.1 5 709
XM_006251084.2 XP_006251146.1 5 943
GenBank mRNA U52095.1 - 5 503
U52102.1 - 5 677

View Full Product Details