Gene Transcripts

Gene Symbol
Trio
Entrez Gene ID
310192
Gene Name
trio Rho guanine nucleotide exchange factor
Gene Aliases
Chromosome Location
Chr. 2: 80471398 - 80769131 on Build Rnor_6.0
UniGene ID
Rn.222545
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003749226.3 XP_003749274.1 9 2044
XM_006232090.2 XP_006232152.1 9 2044
XM_006232091.2 XP_006232153.1 9 1919

View Full Product Details