Gene Transcripts

Gene Symbol
Lztr1
Entrez Gene ID
360745
Gene Name
leucine-zipper-like transcription regulator 1
Gene Aliases
Chromosome Location
Chr. 11: 87381641 - 87397849 on Build Rnor_6.0
UniGene ID
Rn.15759
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108326.2 NP_001101796.1 10 1135
XM_006248707.2 XP_006248769.1 10 1448
GenBank mRNA BC169071.1 - 10 1135
FQ211947.1 - 10 1190

View Full Product Details