Gene Transcripts

Gene Symbol
Olr250
Entrez Gene ID
405923
Gene Name
olfactory receptor 250
Gene Aliases
Chromosome Location
Chr. 1: 172348583 - 172349530 on Build Rnor_6.0
UniGene ID
Rn.145382
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001037.1 NP_001001037.1 1 445

View Full Product Details