Gene Transcripts

Gene Symbol
Vps72
Entrez Gene ID
310661
Gene Name
vacuolar protein sorting 72 homolog (S. cerevisiae)
Gene Aliases
Tcfl1
Chromosome Location
Chr. 2: 196277956 - 196289549 on Build Rnor_6.0
UniGene ID
Rn.1986
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107695.1 NP_001101165.1 5 637
GenBank mRNA BC161999.1 - 5 625

View Full Product Details