Gene Transcripts

Gene Symbol
Otp
Entrez Gene ID
294640
Gene Name
orthopedia homeobox
Gene Aliases
Chromosome Location
Chr. 2: 24546393 - 24554953 on Build Rnor_6.0
UniGene ID
Rn.7202
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100523.1 NP_001093993.1 3 629

View Full Product Details