Gene Transcripts

Gene Symbol
LOC317165
Entrez Gene ID
317165
Gene Name
similar to Set alpha isoform
Gene Aliases
LRRGT00152
Chromosome Location
Chr. 18: 40853988 - 40873685 on Build Rnor_6.0
UniGene ID
Rn.145124
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001047892.1 NP_001041357.1 6 1371
GenBank mRNA AY539903.1 - 6 1371

View Full Product Details