Gene Transcripts

Gene Symbol
Ticam2
Entrez Gene ID
364867
Gene Name
toll-like receptor adaptor molecule 2
Gene Aliases
Chromosome Location
Chr. 18: 40555106 - 40572417 on Build Rnor_6.0
UniGene ID
Rn.225789
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108890.1 NP_001102360.1 3 819
XM_006254690.2 XP_006254752.1 2 892

View Full Product Details