Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001017476.1 | NP_001017476.1 | 6 | 1156 | |
XM_006246777.2 | XP_006246839.1 | 6 | 1303 | ||
XM_006246778.2 | XP_006246840.1 | 6 | 1307 | ||
XM_006246781.2 | XP_006246843.1 | 6 | 1187 | ||
XM_006246782.2 | XP_006246844.1 | 6 | 1300 | ||
GenBank mRNA | BC079326.1 | - | 6 | 1156 |