Gene Transcripts

Gene Symbol
Hus1
Entrez Gene ID
498411
Gene Name
HUS1 checkpoint homolog (S. pombe)
Gene Aliases
Chromosome Location
Chr. 14: 89194086 - 89208825 on Build Rnor_6.0
UniGene ID
Rn.12812
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109092.1 NP_001102562.1 7 786
XM_006251468.1 XP_006251530.1 7 781
XM_006251469.2 XP_006251531.1 7 985

View Full Product Details