Gene Transcripts

Gene Symbol
LOC500948
Entrez Gene ID
500948
Gene Name
LRRGT00073
Gene Aliases
Chromosome Location
Chr. 8: 16279392 - 16280134 on Build Rnor_6.0
UniGene ID
Rn.154987
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001047964.1 NP_001041429.1 568
GenBank mRNA AY387059.1 - 568

View Full Product Details