Gene Transcripts

Gene Symbol
CDH15
Entrez Gene ID
1013
Gene Name
cadherin 15
Gene Aliases
CDH14, CDH3, CDHM, MCAD, MRD3
Chromosome Location
Chr.16: 89171755 - 89195492 on Build GRCh38
Uni Gene
Hs.148090
Interrogated Sequence Translated Protein
RefSeq NM_004933.2 NP_004924.1
GenBank mRNA AK308596.1 -
AK308631.1 -
AW452260.1 -
BC008951.2 -
D83542.1 -
ENST00000289746.0 -

View Full Product Details