Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_000247.2 | NP_000238.1 |
NM_001177519.2 | NP_001170990.1 | |
NM_001289152.1 | NP_001276081.1 | |
NM_001289153.1 | NP_001276082.1 | |
NM_001289154.1 | NP_001276083.1 | |
GenBank mRNA | AB506764.1 | - |
AK094237.1 | - | |
AK225393.1 | - | |
AK299135.1 | - | |
AK310028.1 | - | |
AK311667.1 | - | |
AY204547.1 | - | |
AY750850.1 | - | |
BC016929.1 | - | |
BM552351.1 | - | |
BM991681.1 | - | |
BX342160.2 | - | |
CN483948.1 | - | |
DT219772.1 | - | |
ENST00000364810.0 | - | |
L14848.1 | - |