Gene Transcripts

Gene Symbol
RFPL1S
Entrez Gene ID
10740
Gene Name
RFPL1 antisense RNA 1
Gene Aliases
NCRNA00006, RFPL1-AS, RFPL1-AS1, RFPL1AS
Chromosome Location
Chr. 22: 29437015 - 29442129 on Build GRCh38
UniGene ID
Hs.167750
Species
Homo sapiens
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002727.2
1 1644
GenBank mRNA AJ010230.1 - LIT2045
RFPL1S
1 1644
DA509757.1 -
1 23

View Full Product Details