Gene Transcripts
- Gene Symbol
- ATP13A2
- Entrez Gene ID
- 23400
- Gene Name
- ATPase 13A2
- Gene Aliases
- CLN12, HSA9947, KRPPD, PARK9
- Chromosome Location
- Chr. 1: 16985958 - 17011972 on Build GRCh38
- UniGene ID
- Hs.128866
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY987009.1 |
- |
5 |
645 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AK130020.1 |
- |
|
2017 |
|
Chr. - - [-] |
View Full Product Details