Gene Transcripts
- Gene Symbol
- PP12719
- Entrez Gene ID
- 100653022
- Gene Name
- uncharacterized LOC100653022
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.584854
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF318328.1 |
- |
1 |
348 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
HY100114.1 |
- |
1 |
131 |
|
Chr. 12 - 57818691 - 57819156 [-] |
GenBankmRNA |
AV722292.1 |
- |
1 |
316 |
|
Chr. 12 - 57818589 - 57819301 [-] |
View Full Product Details