Gene Transcripts
- Gene Symbol
- 1700093E11Rik
- Entrez Gene ID
- 67352
- Gene Name
- RIKEN cDNA 1700093E11 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.220761
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK007049.1 |
- |
1 |
74 |
|
AK018923.1 |
- |
1 |
74 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY707119.1 |
- |
1 |
71 |
|
Chr. 2 - 178417473 - 178418240 [-] |
View Full Product Details