Gene Transcripts
- Gene Symbol
- 4930459C07Rik
- Entrez Gene ID
- 74881
- Gene Name
- RIKEN cDNA 4930459C07 gene
- Gene Aliases
- Chromosome Location
- Chr. 10: 96226207 - 96248708 on Build GRCm38
- UniGene ID
- Mm.410819
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_110448.1 |
|
1 |
291 |
|
GenBank mRNA |
AK015487.1 |
- |
1 |
293 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY715035.1 |
- |
1 |
293 |
|
Chr. 10 - 96226207 - 96248350 [+] |
View Full Product Details