Gene Transcripts

Gene Symbol
5730412P04Rik
Entrez Gene ID
70491
Gene Name
RIKEN cDNA 5730412P04 gene
Gene Aliases
Chromosome Location
Chr. X: 136104603 - 136115420 on Build GRCm38
UniGene ID
Mm.396636
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_045424.1 6 953
NR_045425.1 5 598
GenBank mRNA AK017555.1 - 6 955
CB235881.1 - 5 601

View Full Product Details