Gene Transcripts
- Gene Symbol
- LINC01250
- Entrez Gene ID
- 101927554
- Gene Name
- long intergenic non-protein coding RNA 1250
- Gene Aliases
- Chromosome Location
- Chr. 2: 2895048 - 3126026 on Build GRCh38
- UniGene ID
- Hs.638446
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_110228.1 |
|
7 |
2130 |
|
GenBank mRNA |
AK095310.1 |
- |
7 |
2130 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000457478.1 |
- |
7 |
2130 |
|
Chr. 2 - 2895048 - 3126026 [-] |
View Full Product Details