Gene Transcripts
- Gene Symbol
- LOC105371795
- Entrez Gene ID
- 105371795
- Gene Name
- uncharacterized LOC105371795
- Gene Aliases
- Chromosome Location
- Chr. 17: 45168805 - 45171485 on Build GRCh38
- UniGene ID
- Hs.569733
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_135646.1 |
|
2 |
1984 |
|
GenBank mRNA |
AK096785.1 |
- |
2 |
1984 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000589796.2 |
- |
2 |
1984 |
|
Chr. 17 - 45168805 - 45171485 [-] |
View Full Product Details