Gene Transcripts

Gene Symbol
1110028F11Rik
Entrez Gene ID
68690
Gene Name
RIKEN cDNA 1110028F11 gene
Gene Aliases
Chromosome Location
Chr. 11: 87699524 - 87705327 on Build GRCm38
UniGene ID
Mm.342009
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_045139.1 1 1948
GenBank mRNA AK003943.1 - 1 77
BC096659.1 - 1 1957
BY084399.1 - 1 24

View Full Product Details