Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NR_126496.1 | 1 | 670 | ||
NR_126497.1 | 1 | 814 | |||
NR_126498.1 | 1 | 1671 | |||
NR_126500.1 | 1 | 814 | |||
GenBank mRNA | AK090420.1 | - | 1 | 670 | |
AK091232.1 | - | 1 | 1670 | ||
BC128083.1 | - | 1 | 1393 | 40075904 | |
BC144351.1 | - | 1 | 813 | 9052871 |