Gene Transcripts

Gene Symbol
Fam193b
Entrez Gene ID
212483
Gene Name
family with sequence similarity 193, member B
Gene Aliases
IRIZIO, Kiaa1931
Chromosome Location
Chr. 13: 55539316 - 55571120 on Build GRCm38
UniGene ID
Mm.35661
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145382.4 NP_663357.2 8 3245
GenBank mRNA AK040568.1 - 2 1373
AK155237.1 - 8 3239
AK173303.1 - 8 2094
BC021381.1 - 2 835 5043282
BC023015.1 - 2 777 5364758
BC025483.1 - 8 2104 5150988
BC051255.1 - 2 3693

View Full Product Details