Gene Transcripts

Gene Symbol
Oas1h
Entrez Gene ID
246729
Gene Name
2'-5' oligoadenylate synthetase 1H
Gene Aliases
Chromosome Location
Chr. 5: 120861422 - 120873506 on Build GRCm38
UniGene ID
Mm.115351
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159934.1 NP_001153406.1 1 156
NM_145228.2 NP_660263.1 1 156
GenBank mRNA AB067530.1 - 1 156
AK135899.1 - 1 160

View Full Product Details