Gene Transcripts

Gene Symbol
Olfr639
Entrez Gene ID
259088
Gene Name
olfactory receptor 639
Gene Aliases
MOR12-1
Chromosome Location
Chr. 7: 104011750 - 104012700 on Build GRCm38
UniGene ID
Mm.422750
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_147084.1 NP_667295.1 1 585
GenBank mRNA BC145837.1 - 1 635

View Full Product Details