Gene Transcripts

Gene Symbol
Lgi1
Entrez Gene ID
252892
Gene Name
leucine-rich, glioma inactivated 1
Gene Aliases
Chromosome Location
Chr. 1: 256955944 - 256996835 on Build Rnor_6.0
UniGene ID
Rn.83615
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145769.3 NP_665712.1 8 981
GenBank mRNA AJ487517.1 - 8 981
BC089222.1 - 8 1088 7320899

View Full Product Details